Learn More
Invitrogen™ Mouse Urokinase ELISA Kit
For the quantitative detection of Mouse Urokinase
Marque:  Invitrogen™ EEL113
Code produit. 18027464
  • 618.00€ / 96 pièces
Découvrez plus d'offres spéciales
Description
Description
This gene encodes a serine protease involved in degradation of the extracellular matrix and possibly tumor cell migration and proliferation. A specific polymorphism in this gene may be associated with late-onset Alzheimer's disease and also with decreased affinity for fibrin-binding. This protein converts plasminogen to plasmin by specific cleavage of an Arg-Val bond in plasminogen. Plasmin in turn cleaves this protein at a Lys-Ile bond to form a two-chain derivative in which a single disulfide bond connects the amino-terminal A-chain to the catalytically active, carboxy-terminal B-chain. This two-chain derivative is also called HMW-uPA (high molecular weight uPA). HMW-uPA can be further processed into LMW-uPA (low molecular weight uPA) by cleavage of chain A into a short chain A (A1) and an amino-terminal fragment. LMW-uPA is proteolytically active but does not bind to the uPA receptor. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
P06869
9.38 pg/mL
ELISA
Colorimetric Microplate Reader
PLAU
<10%
HRP
RUO
Refer to product documentation for component specific storage temperature.
1 hr 20 min
15.63 to 1000 pg/mL
HRP
Plasma
18792
<10%
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
96 Tests
100 µL
Mouse
3 hr 30 min