Learn More
STEMCELL Technologies Mouse Recombinant MIG (CXCL9)
Click to view available options
Quantité:
25 μg
5 μg
Description
- Monokine induced by interferon-gamma (MIG), or CXCL9, is a member of the CXC chemokine family
- MIG is closely related to two other chemokines: CXCL10 and CXCL11, all of which signal through the CXCR3 receptor (Ding et al.) MIG is secreted by a variety of immune cells including T cells, NK cells, dendritic cells, macrophages, and eosinophils, as well as non-immune cells including hepatic stellate cells, preadipocytes, thyrocytes, endothelial cells, tumor cells, fibroblasts, and glial cells of the central nervous system
- MIG has also been shown to act as a chemoattractant for activated T cells and for tumor-infiltrating leukocytes (TILs), but not for neutrophils or for monocytes
- MIG has also been reported to be both a tumor suppressor and tumor promoter in various types of cancer
Spécification
Spécification
| Nom | Chemokine (C-C-C motif) ligand 9, CMK, crg-10, CXCL9, Gamma interferon-induced monokine, Humig, M119, MIG, Monokine induced by gamma-interferon, SCYB9, Small inducible cytokine B9 |
| Quantité | 5 μg |
| Type de cellule | Monocytes; T Cells |
| Pureté ou qualité | ≥95% |
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.