missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat FABP2/I-FABP Antibody, R&D Systems™
Goat Polyclonal Antibody
Marque: R&D Systems AF1486
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00970kg
Description
FABP2/I-FABP Polyclonal specifically detects FABP2/I-FABP in Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Spécification
FABP2/I-FABP | |
Polyclonal | |
Unconjugated | |
P02693 | |
FABP2 | |
E. coli-derived recombinant rat FABP2/I-FABP Met1-Glu132 Accession # P02693 | |
100 μg | |
Primary | |
Detects mouse and rat FABP2/I-FABP in Western blots. Detects rat FABP2/I-FABP in direct ELISAs. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant rat FABP1, recombinant human FABP3, recombinant mouse (rm) FABP4, and rmFABP5 is observed. | |
Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein | |
Goat | |
Affinity Purified | |
RUO | |
2169 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |