missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GFR alpha-like Antibody, R&D Systems™
Sheep Polyclonal Antibody
Marque: R&D Systems AF5728
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GFR alpha-like Polyclonal specifically detects GFR alpha-like in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Spécification
GFR alpha-like | |
Polyclonal | |
Unconjugated | |
Q6SJE0 | |
Sheep | |
Affinity Purified | |
RUO | |
404194 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Gfral | |
Chinese hamster ovary cell line CHO-derived recombinant mouse GFR alpha-like isoform A Gln20-Gly349 Accession # Q6SJE0 | |
100 μg | |
Primary | |
Detects mouse GFR alpha-like in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu