missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD63 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Marque: R&D Systems MAB5417
396.00 EUR valable jusqu'au 2025-03-28
Utilisez le code promo "25306" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD63 Monoclonal specifically detects CD63 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Spécification
CD63 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/mL, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
P41731 | |
CD63 | |
HEK293 human embryonic kidney cell line transfected with mouse CD63 Met1-Met238 Accession # P41731 | |
100 μg | |
Primary | |
Detects mouse CD63 in direct ELISAs. | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, CyTOF | |
446703 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, Lamp-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30, TSPAN30granulophysin | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
967 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Mouse CD63 Antibody, R&D Systems™ > 100ug; Unlabeled
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu