missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse BMP-6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Marque: R&D Systems MAB6325
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
BMP-6 Monoclonal specifically detects BMP-6 in Mouse samples. It is validated for Neutralization.
Spécification
| BMP-6 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 1.5-7.5 ug/mL | |
| BMP6, BMP-6, bone morphogenetic protein 6, vegetal related growth factor (TGFB-related), vegetal-related (TGFB related) cytokine, VG-1-R, VG-1-related protein, Vg1-related sequence, VGR, Vgr-1, VGR1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse BMP-6 in ELISAs. | |
| Mouse | |
| Purified |
| Neutralization | |
| 719040 | |
| Unconjugated | |
| P20722 | |
| BMP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse BMP-6 Met1-His510 Accession # P20722 | |
| 100 μg | |
| 654 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu