Learn More
Thermo Scientific™ Mixtainer BioProcess Container (BPC) for HyPerforma™ Mixtainer™ Systems
Code produit. p-200078642 Tous les produits Thermo Scientific Produits
Click to view available options
Capacité (métrique):
50 l
100 l
200 l
Détails du port:
4 ports et tubes montés sur le dessus
Port à poudre
Conditionnement:
15 pièces
3 pièces
5 pièces
Code produit. 30040271
Marque: Thermo Scientific™ SH30661.01
The complete Thermo Scientific™ Mixtainer™ System is an integrated, single-use sterile unit for mixing of cell culture media, and other process liquids such as buffers, reagents and bulk drug precursors or products.
Recommended for:
  • Hydration of powdered media and buffers
  • Sterile mixing of cell culture media with sera and reagents
  • Sterile mixing of buffer solutions or other process fluids
  • Pooling of sterile liquid fractions
TRUSTED_SUSTAINABILITY
Spécification
Capacité (métrique) 50 l
Détails du port 4 ports et tubes montés sur le dessus
Tubulure 1 0.25 in. ID Tubing and male/female Luer connectors (Port 1)
Tubulure 2 0.25 in. ID Tubing and male/female Luer connectors (Port 2)
Tubulure 3 3/8 in. ID tubing and quick-connect couplings (Port 3)
Tubulure 4 3/8 in. ID tubing and quick-connect couplings (Port 4)
Tubing Style TPE
Stérilité Sterile