missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17090-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognizes the endogenous MFF protein around 26-29 kDa and 35-38 kDa.
The exact function of C2orf33 remains unknown.Spécification
MFF | |
Polyclonal | |
Unconjugated | |
MFF | |
C2orf33, MFF, mitochondrial fission factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301563, 56947, 75734 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4KM98, Q6PCP5, Q9GZY8 | |
Mff | |
MFF Fusion Protein Ag9990 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |