missing translation for 'onlineSavingsMsg'
Learn More

Melanophilin Mouse anti-Human, Porcine, Clone: 1A8D12, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigène Melanophilin
Clone 1A8D12
Concentration 0.77 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
16837655
View Documents Promotion Details
Proteintech
66092-1-IG-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16827655
View Documents Promotion Details
Proteintech
66092-1-IG-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
Specifications

Specifications

Melanophilin
0.77 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Exophilin 3, melanophilin, MLPH, SLAC2 A, SLAC2A, Synaptotagmin like protein 2a
MLPH
IgG1
Protein G
Antibody
1A8D12
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Porcine, Human
Q9BV36
100049680, 79083
Melanophilin Fusion Protein Ag19286
Primary
-20°C
MLPH
Videos
SDS
Documents

Documents

Product Certifications