Learn More
Macherey-Nagel™ Med-Test Combi 10™ VET, Refill Pack
Tous les produits Macherey-Nagel Produits
Click to view available options
Quantité:
100 barrettes
Conditionnement:
1 pièce
Description
- The use of urine test strips is acknowledged as a modern screening method in medical practice.
- Within minutes health status can be obtained.
- Simplifies the decision about further diagnostic and therapeutic action.
Spécification
Spécification
| Format | Urine Test Strip |
| Type | Urinalysis Test: Glucose, Ketones, Protein, Blood, Leukocytes,Nitrite, Specific Gravity, pH, Bilirubin and Urobilinogen |
| Type d’emballage | Refill Pack |
| Quantité | 100 barrettes |