Learn More
TFA Dostmann Mechanical Stopwatch
Click to view available options
Plage de durée:
0 à 30 min
Conditionnement:
1 pièce
Beskrivning
This mechanical stopwatch made of metal is particularly sturdy and always ready for use. It is traditionally used for timekeeping in training or competition, but it can also be used in many other areas, such as industry, crafts and laboratories. The stopwatch is wound up by hand and then runs for up to 30 minutes. After the stop, you can read the time to 1/5 second. Carry bag and neck strap are included.
- Start/stop with crown
- Lateral reset push button
- Time-out possible
- Wind-up mechanism
- 13 jewels
- Chromed metal case
- 1/5 second increments
- Including neck strap and bag
Specifikationer
Specifikationer
| Type de produit | Analog Stopclock |
| Plage de durée | 0 à 30 min |
| Poids (métrique) | 98 g |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering