Learn More
Bolle Safety X5 Masterall dual lens goggle BOLLE
Double lens and anti-fog technology enable increased protection.
Marque:  Bolle Safety MASTEDEPSI
Code produit. 15528291
Description
Description
  • Equalizer technology filters and regulates moisture levels in goggles
  • Permanent scratch-resistant platinum coating
  • Sealed PVC frame and neoprene strap
  • For use in offshore and petrochemical environments
Documents
One moment while we fetch your results.
Nom du produit
Bolle Safety X5 Masterall dual lens goggle BOLLE > Double lens