missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP3K6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16212-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK6 activates the JNK, but not ERK or p38 kinase pathways.Spécification
MAP3K6 | |
Polyclonal | |
Unconjugated | |
MAP3K6 | |
ASK2, MAP3K6, MAPKKK6, MEKK6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
313022, 53608, 9064 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95382, Q9WTR2 | |
Map3k6 | |
MAP3K6 Fusion Protein Ag9206 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |