Learn More
LTBP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA551930
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: ERSPNLRRSS AAGEGTLARA QPPAPQSPPA PQSPPAGTLS GLSQTHPSQQ HVGLSRTVRL HPTATASSQL SSNALPPGPG LEQRDGTQQA VPLEHPSSPW GLNLTEKIKK IKIVFTPTIC KQTCARGHCA NSCERG Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
The protein encoded by this gene belongs to the family of latent transforming growth factor (TGF)-beta binding proteins (LTBP), which are extracellular matrix proteins with multi-domain structure. This protein is the largest member of the LTBP family possessing unique regions and with most similarity to the fibrillins. It has thus been suggested that it may have multiple functions: as a member of the TGF-beta latent complex, as a structural component of microfibrils, and a role in cell adhesion.Spécification
LTBP2 | |
Polyclonal | |
Unconjugated | |
LTBP2 | |
AW208642; C14orf141; GLC3D; latent transforming growth factor beta binding protein 2; latent-transforming growth factor beta-binding protein 2; LTBP2; LTBP-2; LTBP3; MSPKA; MSTP031; WMS3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4053 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14767 | |
LTBP2 | |
Recombinant protein corresponding to Human LTBP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only