Learn More
LRRC43 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA565236
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Immunogen sequence: SELSVISGPST ILQMPRASAE ELAKLRLRID PRLCPSPGTV LFSTAHKPWA EVIPCSYEMQ HSLRDLVPLK AFLLAGTTVT IVEEKILSWP Highest antigen sequence identity to the following orthologs - mouse 65%, rat 66%.
LRRC43 is a protein coding gene.Spécification
LRRC43 | |
Polyclonal | |
Unconjugated | |
LRRC43 | |
leucine rich repeat containing 43; leucine-rich repeat-containing protein 43; LRRC43 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N309 | |
LRRC43 | |
Recombinant Human LRRC43 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only