Learn More
Lenz Laborglasintrumente Long Ground Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marque: Lenz Laborglasintrumente 01100719
Code nomenclature Nacres: NB.43
Informations supplémentaires : Poids : 0.25000kg
Description
- Cônes rodés coniques
- En tubulure DURAN™
Spécification
NS 19/38 | |
Verre borosilicaté 3.3 |