Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points Traceable Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Marque: Brannan™ 44/809/8 + 49/101/0
Code nomenclature Nacres: PB.12
Informations supplémentaires : Poids : 0.02400kg
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Spécification
0°C, 20°C, 45°C | |
Total Immersion | |
Glass | |
122°F | |
-31°F | |
-31°F to +122°F | |
± 1.0°C at 35°C | |
1.2 cm |
ISO 17025 UKAS | |
30.5 cm | |
Blue Spirit | |
+50°C | |
-35°C | |
-35°C to +50°C | |
0.5°C |