Learn More
Brannan™ LO-tox™ Lab Thermometer with 3-points UKAS Calibration
LO-tox™ lab thermometer complete with calibration certificate.
Marque: Brannan™ 44/822/0 + 49/503/0
Code nomenclature Nacres: PB.12
Informations supplémentaires : Poids : 0.03900kg
Description
- LO-tox™ is an organic filling specially chosen to achieve a wide temperature span and provide the lowest practical levels of hazard
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Spécification
0°C, 25°C, 45°C | |
Partial Immersion | |
Glass | |
124°F | |
30°F | |
30°F to 124°F | |
± 1.0°C at 35°C | |
76 mm |
ISO 17025 UKAS | |
40.5 cm | |
Blue Spirit | |
+51°C | |
-1°C | |
-1°C to 51°C | |
0.2°C | |
1.2 cm |