Learn More
Gilac™ Lid for 12 L Round Bucket with Pouring Spout
Lid for 12 L round bucket with pouring spout.
Marque: Gilac™ G615451
Code nomenclature Nacres: NB.51
Informations supplémentaires : Poids : 0.24000kg
Description
- Lid for 12 L round bucket with pouring spout.
Spécification
Stainless Steel | |
Lid | |
1 Ea. |