missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP1 Antibody (RL13) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NB120-2737-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
LAP1 Monoclonal specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Spécification
| LAP1 | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot 1:200, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| TOR1AIP1 | |
| Pore complex-lamina fraction isolated from rat liver nuclear envelopes. | |
| 50 μL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 208263 | |
| Mouse | |
| Purified |
| Western Blot | |
| RL13 | |
| Unconjugated | |
| lamina-associated polypeptide 1B, torsin A interacting protein 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat tissues. This does not cross-react with LAP 2.This detects 3 distinct proteins of molecular weights 75, 68 and 55 kDa representing LAP 1A, 1B and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with NB120-2737 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining. | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu