Learn More
Thermo Scientific™ LanthaScreen™ Tb-p-c-jun (pSer73) Antibody
For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications.
Marque: Thermo Scientific™ PV4449
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00900kg
Description
- A part of the activating protein-1 (AP-1) transcription factor, c-Jun is phosphorylated at Ser73 by members of the JNK family
- For use in combination with the GFP-c-Jun physiological protein substrate (PV4446) to evaluate inhibitors in medium to high throughput screening applications
Spécification
Tb (Terbium)/Lanthanide Chelates | |
Terbium-labeled p-c-Jun (pSer73) antibody in HEPES buffered saline. Store at -20°C. | |
Human | |
End Point | |
Ser/Thr Kinases |
25 μg | |
340/490, 520 | |
Dry Ice | |
Hu | |
LanthaScreen™ |
For Research Use Only. Not for use in diagnostic procedures.