Learn More
Labplas Sani-Sponge Sampling Bags, Regular tabs: With Clear Body, Tear-Off Protection Strip
Click to view available options
Comprend:
Tampon neutralisant DE
Unit Size:
100 pièces
Description
- Detects Listeria, Salmonella and E. coli
- Contaminant-free
- Virgin polyethylene
- Large bag mouth opening
- Easy to use pull tabs
- Sponge for swabbing
- Dehydrated sponge
Specifications
Specifications
| Description | DE Neutralizing buffer |
| Comprend | Tampon neutralisant DE |
| Volume (métrique) | 450 mL |
| Matériau | Polyethylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction