Learn More
L3MBTL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA528549
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
Recommended positive controls: NT2D1, mouse testis. Predicted reactivity: Mouse (88%), Rat (87%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
L3MBTL2 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Spécification
L3MBTL2 | |
Polyclonal | |
Unconjugated | |
L3MBTL2 | |
4732493N06Rik; H-l(3)mbt-l; H-l(3)mbt-like protein 2; l(3)mbt-like 2; l(3)mbt-like 2 (Drosophila); l(3)mbt-like 2 protein; L(3)mbt-like protein 2; L3MBT; L3MBTL histone methyl-lysine binding protein 2; L3MBTL2; L3MBTL2 polycomb repressive complex 1 subunit; L3MBTL2, polycomb repressive complex 1 subunit; lethal(3)malignant brain tumor-like 2 protein; lethal(3)malignant brain tumor-like protein 2; lmbt-like 2 (Drosophila) isoform 1; lmbt-like 2 (Drosophila) isoform 2; M4mbt | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214669, 83746 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P59178, Q969R5 | |
L3MBTL2 | |
Recombinant fragment corresponding to a region within amino acids 406 and 705 of Human L3MBTL2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |