Learn More
Melford L-Arginine HCl
Click to view available options
Quantité:
100 g
500 g
1 kg
5 kg
Conditionnement:
100g
1kg
500g
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 1119-34-2
- Molecular Formula: C6H14N4O2 HCl
- Molecular Weight: 210.7
Spécification
Spécification
| Nom chimique ou matériau | L-Arginine HCl |
| CAS | 1119-34-2 |
| Formule moléculaire | C6H14N4O2-HCl |
| Quantité | 1 kg |