missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv4.1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13768-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The KV gene family encodes more than 30 genes that comprise the subunits of the K+ channels, and they vary in their gating and permeation properties, subcellular distribution and expression patterns. Functional KV channels assemble as tetramers consisting of pore-forming alpha-subunits (KV alpha), which include the KV1, KV2, KV3 and KV4 proteins, and accessory or KV beta subunits that modify the gating properties of the coexpressed KV alpha subunits. Differences exist in the patterns of trafficking, biosynthetic processing and surface expression of the major KV1 subunits (KV1.1, KV1.2 and KV1.4) expressed in rat and human brain, suggesting that the individual protein subunits are highly regulated to control for the assembly and formation of functional neuronal channels.Spécification
Kv4.1 | |
Polyclonal | |
Unconjugated | |
KCND1 | |
KCND1, Kv4.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116695, 16506, 3750 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03719, Q9NSA2 | |
Kcnd1 | |
Kv4.1 Fusion Protein Ag4751 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |