missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRT5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marke: Abnova™
Code nomenclature Nacres: NA.55
Weitere Details : Poids : 0.00010kg
Spezifikation
Western Blot | |
KRT5 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 62.4kDa | |
Transient overexpression cell lysate was tested with anti-KRT5 antibody (Mfr. No. H00003852-D01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC024292.1 | |
pCMV-KRT5 full-length | |
100 μL | |
Human |
Sicherheit und Handhabung
Signalwort
- Attention