missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL25 Antibody, HRP, Novus Biologicals™
Chicken Polyclonal Antibody
Marque: Novus Biologicals NBP1-77147H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
KLHL25 Polyclonal antibody specifically detects KLHL25 in Human samples. It is validated for Western Blot, ELISASpécification
KLHL25 | |
Polyclonal | |
PBS | |
Chicken | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
HRP | |
ectodermal-neural cortex 2, ENC-2BTB/POZ KELCH domain protein, ENC2ectoderm-neural cortex protein 2, FLJ12587, FLJ45015, kelch-like 25 (Drosophila), Kelch-like protein 25 | |
Antibody was raised against a 14 amino acid synthetic peptide near the center of human ENC-2. The immunogen is located within amino acids 230 - 280 of ENC-2. Amino Acid Squence: SSEALMADERTKL | |
0.1 mL | |
Neuroscience | |
64410 | |
Store at 4°C in the dark. | |
IgY |