missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14883-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Serves as a substrate-specific adapter for the CUL3-based ubiquitin-protein E3 ligase complex. Negatively regulates the Wnt signaling pathway via the targeted ubiquitination and subsequent proteolysis of DVL3.Tissue specificity: Highly expressed in testis and at lower levels in the submandibular salivary gland.Spécification
KLHL12 | |
Polyclonal | |
Unconjugated | |
KLHL12 | |
C3IP1, CUL3 interacting protein 1, DKIR, kelch like 12 (Drosophila), Kelch like protein 12, KLHL12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
240756, 266772, 59349 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53G59, Q8BZM0, Q8R2H4 | |
KLHL12 | |
KLHL12 Fusion Protein Ag6658 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |