Learn More
BUCHI Kjeldahl Sample Tubes
Sample tubes for standard Kjeldahl are suitable for most applications.
Spécification
Type Tube à échantillons
Consulter d'autres spécifications
Description
Description
  • Precision joint: tightness, avoid fume leaks
  • Constricted condensation zone; no splashing
  • Wider diameter; reduced foaming
  • Thicker glass wall, longer lifetime
Spécification
Spécification
Tube à échantillons