missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR2DL4 Monoclonal Antibody (mAb#33), PE, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA110111
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody recognizes the extracellular portion of CD158d.
CD158d / KIR2DL4 is a KIR family member that shares structural features with both activating and inhibitory receptors and may mediate different functions under different circumstances. It contains cytoplasmic ITIM, suggesting inhibitory function, but also transmembrane domain similar to those of activating KIRs. It has been reported that CD158d serves as an inhibitory receptor for peripheral and uterine NK cells, but its ligation with soluble mAbs (unlike immobilized mAbs) results in activation of IFN-gamma secretion. CD158d also binds both membrane form and soluble form of its ligand HLA-G.Spécification
KIR2DL4 | |
Monoclonal | |
PE | |
KIR2DL4 | |
CD158 antigen-like family member D; CD158D; G9P; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 4; killer cell immunoglobulin-like receptor 2DL4; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4; killer cell inhibitory receptor 103AS; killer Ig receptor; killer-cell immunoglobulin-like receptor; killer-cell immunoglobulin-like receptor 2DL4; KIR; KIR103; KIR103AS; KIR-103AS; KIR2DL4; KIR-2DL4; MHC class I NK cell receptor KIR103AS; natural killer cell inhibitory receptor; natural killer cell receptor; NK cell receptor | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
3805 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
mAb#33 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q99706 | |
KIR2DL4 | |
NK3.3 cells and KIR2DL4-Ig fusion protein | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu