missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble™ KIMAX™ Automatic Burets with Bottles
Class A
1030.00€
Spécification
Matériau | Verre borosilicaté |
---|---|
Type | Burette automatique |
Code produit | Marque | Capacité (métrique) | Graduations | Joint conique standard | Prix | Quantité et disponibilité | |||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Capacité (métrique) | Graduations | Joint conique standard | Prix | Quantité et disponibilité | |||
17798924
|
DWK Life Sciences
17124F-10 |
10 mL | 0,05 mL | 29 |
1030.00€
1 pièce |
Expédition estimée: 03-10-2024
Connectez-vous pour voir le stock disponible |
|||
Description
standard taper joints fit into reagent bottles with standard taper necks.
- Constructed with overflow tubes that return directly to reservoir
- For air intake, overflow cups have hose connection for attaching drying tube; second drying tube provided for use between rubber bulb and reservoir
- Precision bore tubing permanently marked with individual serial number
- Durable black ceramic enamel scale
- Inset shows top configuration
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST™.
Spécification
Verre borosilicaté | |
Burette automatique |