missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunB Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA519135
357.00 EUR valable jusqu'au 2025-03-16
Utilisez le code promo "24070" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
JunB, a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.
Spécification
JunB | |
Polyclonal | |
Unconjugated | |
Junb | |
activator protein 1; AP-1; Jun B; jun B proto-oncogene; Junb; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; myD21; Oncogene Jun-B; Transcription factor jun-B | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3726 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P17275 | |
Junb | |
Synthetic peptide sequence (TLKAENAGLSSTAG) corresponding to the internal amino acids of JUNB | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
JunB Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu