Learn More
Brady™ ISO Safety Sign - Disinfect Surface, Dutch
Sign to signify that the surface must be disinfected. Text is in Dutch.
Marque: Brady™ 323319
Description
- Sign compliant to ISO 7010
Spécification
ISO 7010, Compliant with Directive 2011/65/EU and 2015/863 | |
M062 | |
1 Ea. | |
Facility Identification, Safety Identification | |
297 mm | |
8.27 in. | |
-40°C to +120°C | |
-40°F to +248°F | |
210 x 297 mm | |
Personal Hygiene Sign |
M/M062/NL540/PP-210x297-1 | |
Polypropylene | |
Surface Mount, Wall Mount | |
11.69 in. | |
1.55 mm | |
210 mm | |
Rectangular | |
Dutch | |
Oppervlak desinfecteren | |
White on Blue |
Suggestions de produits
Customers who viewed this item also viewed.