Learn More
Brady™ ISO Safety Sign - Disinfect Surface, Dutch
Sign to signify that the surface must be disinfected. Text is in Dutch.
Marque:  Brady™ 323324
Code produit. 30280519
  • 25.55€ / 1 pièce
Expédition estimée: 06-05-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Description
  • Sign compliant to ISO 7010
Spécification
Spécification
ISO 7010, Compliant with Directive 2011/65/EU and 2015/863
M062
1 Ea.
Facility Identification, Safety Identification
210 mm
5.83 in.
-40°C to +140°C
-40°F to +284°F
148 x 210 mm
Personal Hygiene Sign
M/M062/NL540/ALU-148x210-1
Aluminum
Surface Mount, Wall Mount
8.27 in.
1 mm
148 mm
Rectangular
Dutch
Oppervlak desinfecteren
White on Blue
Nom du produit
Brady™ ISO Safety Sign - Disinfect Surface, Dutch > Aluminium, Language: Dutch, 148 x 210 mm