Learn More
Brady™ ISO Safety Sign - Disinfect Surface, Dutch
Sign to signify that the surface must be disinfected. Text is in Dutch.
Marque: Brady™ 323308
Description
- Sign compliant to ISO 7010
Spécification
ISO 7010, Compliant with Directive 2011/65/EU and 2015/863 | |
Gloss | |
Laminated Polyester | |
Surface Mount, Wall Mount | |
4.13 in. | |
0.1 mm | |
297 mm | |
Rectangular | |
Dutch | |
Oppervlak desinfecteren | |
White on Blue |
M/M062/NL540/PE-297x105-1 | |
M062 | |
1 Ea. | |
Facility Identification, Safety Identification | |
105 mm | |
11.69 in. | |
-40°C to +110°C | |
-40°F to +230°F | |
297 x 105 mm | |
Personal Hygiene Sign |
Suggestions de produits
Customers who viewed this item also viewed.