missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ipilimumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Marque: Invitrogen MA541799
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ipilimumab (Yervoy) is a humanized monoclonal antibody that is approved by the U.S. Food and Drug Administration for the treatment of patients with unresectable or metastatic melanoma. Ipilimumab works to activate the immune system by targeting CTLA-4 (Cytotoxic T-lymphocyte protein 4), a negative regulator of cytotoxic T-lymphocyte activity. Ipilimumab binds to CTLA-4, blocking the inhibitory signal, which allows the cytotoxic T-lymphocytes to destroy the cancer cells.Spécification
Ipilimumab | |
Recombinant Monoclonal | |
Unconjugated | |
BMS-734016; MDX-010; MDX-CTLA-4; Yervoy | |
Human CTLA4/CD152 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
2.87 mg/mL | |
PBS with no preservative; pH 7.5 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ |