Learn More
B Braun™ Introcan Safety™ Safety Vein Indwelling Cannula
Reduce needlestick injuries and infection. B Braun™ Introcan Safety™ Safety Vein Indwelling Cannula is ideal for reducing needlestick injuries and associated infection. The cannula has a fixation wing secures the needle tip immediately after pulling out.
Marque: B Braun™ 4254503-01
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Safety venous indwelling cannula with FEP or polyurethane catheter for a venous gentle placement and uncomplicated residence time
- Handle design for one-hand puncture technique
- Excellent puncture properties due to atraumatic cannula tip with 3-facet cut geometry
- Highly transparent blood chamber
- Capillary: Four fully embedded X-ray contrast strips
- Hydrophobic blood catcher plugs
- Removable lock closure cone
- Lock approach
- Color size indicator
- Latex-free
- PVC-free
- According to ISO standard 10.555-1 / 5
- Self-activating clip technique
- Safety-Clip secures the needle tip immediately after pulling out
- Safety mechanism can not be bypassed
- Minor customization of user habits
Spécification
FEP | |
PVC-free | |
0.70 mm |
Jaune | |
24 G | |
No |