Learn More
Cole-Parmer™ Accessoires pour blocs chauffants Integrity Code produit.: 12709048
Ajouter au panier
Cole-Parmer™ Accessoires pour blocs chauffants Integrity
Code produit. 12709048
Capteur IR non intrusif
Click to view available options
Description:
Boîte IR multiple
Capteur IR non intrusif
Logiciel d’intégrité
Module multitempérature
PID-NIR5-BNSD
Tubes en verre, 24 mm x 150 mm
Code produit. 12709048
Marque: Cole-Parmer™ 3663090
Ajouter au panier
Ajouter au panier
Keep your Integrity 10, RS-200, or RS-250 Reaction Station running smoothly with these parts and accessories.
  • Suitable for use with Cole-Parmer™ Integrity 10, RS-200, or RS-250 series reaction stations.
  • Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Spécification
Description Capteur IR non intrusif
À utiliser avec (équipement) Integrity 10
Type Accessoire, capteur IR
  • Electrothermal - Integrity 10 reaction station
    The STEM Integrity 10 reaction station enables you to conduct 10 different reactions simultaneously within the same reaction unit, each reaction being conducted within its own cell, at its own individual temperature and stir rate.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • subtitles off, selected
      Electrothermal Integrity 10 reaction station
    Correction du contenu d'un produit
    Nom du produit
    Cole-Parmer™ Accessoires pour blocs chauffants Integrity >