missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ILK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | ILK |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16803445
|
Proteintech
24926-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16893435
|
Proteintech
24926-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. ILK is a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction.Spécification
ILK | |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O55222, Q13418 | |
16202, 3611 | |
ILK Fusion Protein Ag21494 | |
Primary | |
-20°C | |
ILK |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ILK, ILK 1, ILK 2, ILK1, ILK2, integrin linked kinase, Integrin linked protein kinase, P59, p59ILK | |
ILK | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |