Learn More
Invitrogen™ IL-18 Human Matched Antibody Pair
Matched Antibody Pair
Marque:  Invitrogen™ BMS267-2MST
Code nomenclature Nacres: NA.41
Code produit. 15511857
  • 1064.00€ / 10 pièces
Includes:
  • 1 vial (1.1mL) monoclonal Coating Antibody to human IL-18 (100μg/mL)
  • 1 vial (55μL) Biotin-Conjugate anti-human IL-18 monoclonal antibody
  • 1 vial (22μL) Streptavidin-HRP
  • 1 vial human IL-18 Standard protein lyophilized, 100ng/mL upon reconstitution
  • 2 vials (50mL) Sample Diluent
Description
Description
The Human IL-18 Matched Antibody Pair Kit comes with pre-matched antibody pairs,standards,sample diluent and streptavidin-HRP to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-18. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production,and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine,and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
IL-18
Monoclonal
Q14116
IL18
RUO
3606
-20°C
ELISA
Biotin
Interleukin-18, Interferon-gamma-inducing factor
10 x 96 Tests
Primary
Human
ELISA Kit