missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Monoclonal Antibody (72C627), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA141046
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Suggested positive control: HeLa or NIH-3T3 whole cell lysate.
IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.Spécification
IKK gamma | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
O88522, Q9Y6K9 | |
Ikbkg | |
His-tagged full-length human IKK3/IKKg. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
72C627 | |
Unconjugated | |
Ikbkg | |
1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9 | |
Mouse | |
Protein G | |
RUO | |
16151, 8517 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu