Learn More
IgG light chain (Kappa) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14678-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16853434
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
This antibody detects the light chain (Kappa) of human IgG.
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.
Spécification
Spécification
IgG light chain (Kappa)
Polyclonal
Unconjugated
IGK
IGK, IGK-, IGKC, immunoglobulin kappa locus
Rabbit
Antigen Affinity Chromatography
RUO
243469, 50802
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P0DOX7
IGK
IgG light chain (Kappa) Fusion Protein Ag6389
150 μL
Primary
Human, Mouse