missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG light chain (Kappa) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | IgG light chain (Kappa) |
---|---|
Concentration | 0.32 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16859083
|
Proteintech
11781-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16849083
|
Proteintech
11781-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.Spécification
IgG light chain (Kappa) | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P01602 | |
28299 | |
IgG light chain (Kappa) Fusion Protein Ag2406 | |
Primary | |
-20°C | |
IGKV1-5 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IGKV, IGKV1 5, IGKV15, IGKV1-5, L12, L12a, V1 | |
IGKV1-5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |