missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGF1R alpha Monoclonal Antibody (24-57), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA513812
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA5-13812 targets IGF-1 Receptor alpha in IA and IP applications and shows reactivity with Human and Rabbit samples. The MA5-13812 immunogen is iGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.
IGF1R (Insulin-like Growth Factor1 Receptor) is a receptor tyrosine kinase that mediates actions of insulin-like growth factor 1 (IFG1). It binds IGF1 with high affinity and IGF2/insulin with lower affinity. Activated IGF1R is involved in cell growth and survival control. IGF1R is crucial for tumor transformation and survial of malignant cells. Ligand binding activates the receptor kinase and leads to receptor autophosphorylation and tyrosines phosphorylation of multiple substrates that function as signaling adapter proteins.Spécification
IGF1R alpha | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P08069 | |
Igf1r | |
IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA | |
500 μL | |
Primary | |
Human, Rabbit | |
Antibody | |
IgG1 κ |
Immunoprecipitation, Inhibition Assays | |
24-57 | |
Unconjugated | |
Igf1r | |
A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; Igf1r; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; insulin-like growth factor 1 receptor beta subunit; insulin-like growth factor I receptor; insulin-like growth factor receptor 1; insulin-like growth factor-I receptor; insulin-like growth factor-I receptor precursor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
Protein G | |
RUO | |
100144342, 3480 | |
4° C | |
Liquid |