Learn More
IGF1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28530-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The IGF1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human IGF1. This antibody recognizes human antigen. The IGF1 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
IGF1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
3479 | |
IgG |
Western Blot, Immunohistochemistry | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |