missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-1 Mouse anti-Human, Clone: 2F9A8, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | ICAM-1 |
---|---|
Clone | 2F9A8 |
Concentration | 0.7 mg/mL |
Applications | Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16807385
|
Proteintech
60299-1-IG-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16897375
|
Proteintech
60299-1-IG-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Intercellular adhesion molecule-1 (ICAM-1, CD54) is an integral membrane protein, a member of the immunoglobulin superfamily, and a ligand for lymphocyte function-associated (LFA) antigens, a beta 2 leukocyte integrin. The normal function of human ICAM-1 is to provide adhesion between endothelial cells and leukocytes after injury or stress. ICAM-1 binds to leukocyte function-associated antigen (LFA-1) or macrophage-1 antigen (Mac-1). It is found on leukocytes, fibroblasts, epithelial cells and endothelial cells and its expression is regulated by inflammatory cytokines. ICAM-1 has a tissue distribution similar to that of the major histocompatibility complex class II antigens and is likely to play a role in inflammatory responses.Spécification
ICAM-1 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BB2, CD54, ICAM 1, ICAM1, ICAM-1, P3.58 | |
ICAM1 | |
IgG2b | |
Protein A | |
Antibody |
2F9A8 | |
Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P05362 | |
3383 | |
ICAM-1 Fusion Protein Ag8309 | |
Primary | |
-20°C | |
ICAM1 |