Learn More
Microbiologics™ Brevundimonas diminuta ATCC® 19146™†
For culture purposes and QC applications
Marque: Microbiologics™ 0805P
Code nomenclature Nacres: NA.7
Informations supplémentaires : Poids : 0.80000kg Transport : UN number : 3373 Chem class : 6.2
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single strain of Brevundimonas diminuta in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2
- The most economical reference stock culture option; lyophilized pellet containing a single strain of Brevundimonas diminuta
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 pellets
- ATCC™ 19146™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Spécification
Brevundimonas diminuta | |
1 | |
2°C to 8°C | |
Kwik-stik™ | |
Culture purpose and QC application |
19146 | |
CE, online certificate of analysis | |
Lyophilized Pellet | |
KWIK-STIK | |
2 U |