Learn More
Juchheim Laborgeraete™ Support élévateur hydraulique de laboratoire
Code produit. 10204782
Click to view available options
Hauteur (métrique):
625 mm
1 060 mm
Conditionnement:
1 pièce
Code produit. 10204782
Marque: Juchheim Laborgeraete™ 02296
Il en reste null
Ajouter au panier
Ajouter au panier
A laboratory lifting table with smooth-running lifting hydraulics that loads up to 85 kg.
  • For static and dynamic loads
  • Particularly stable due to scissor construction made of robust steel
  • Acid and base resistant due to high-quality powder coating
  • Requires no external connections (pump hydraulics fully integrated)
  • With handles for easy transport
  • To lower the load, the lever only has to be pressed slightly upwards. The load then sinks slowly (approx. 3 cm/s) and stops as soon as the lever is released again.
Spécification
Matériau Acier
Hauteur (métrique) 1 060 mm
Description Adjustment Range: 230 to 1060 mm