missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human Transferrin protein
HumanKine™ recombinant protein
Marque: Proteintech HZ-1317-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Transferrin is an 80kDA, monomeric glycoprotein that is responsible for binding ferric iron and transporting it throughout the body. It is primarily produced by hepatocytes and is a critical component of iron homeostasis as it delivers free iron to tissue sites where it is absorbed and utilized by the body. Once it reaches a site of absorption, iron-loaded transferrin binds to its corresponding transferrin receptor and taken up by the cell through endocytosis, after which the iron is disassociated, and free transferrin is released back into the bloodstream at a turnover rate of around 10 times per day. In addition to iron homeostasis, transferrin also plays an important role in the innate immune system. Downregulation of transferrin receptor expression has been linked to the reduced ability of intracellular pathogens to access a cell’s iron supply. Measurement of transferrin levels in the blood can also serve as a clinical marker for possible iron deficiency (PMID: 30422523, 22033148).Spécification
P02787 | |
7018 | |
100 μg | |
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) | |
Human | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
>95% |
1 x PBS | |
70 kDa | |
HEK293 | |
<0.1 EU/μg | |
Recombinant | |
Lyophilized |