Learn More
Proteintech™ HumanKine™ Recombinant Human Noggin Protein
Code produit. 16841326 Tous les produits Proteintech Produits
Click to view available options
Quantité:
10 μg
100 μg
1 000 μg
Conditionnement:
1000µg
100µg
10µg
Code produit. 16841326
Marque: Proteintech™ HZ11181000UG
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Spécification
Identification génétique (Entrez) 9241
Poids moléculaire 65 kDa, homodimer, glycosylated
Quantité 1 000 μg
Espèces Human
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Contenu et stockage Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Activité Typically ≤ 15 ng/mL EC50
Système d’expression Human embryonic kidney 293 (HEK293) human cell expression system
Forme Lyophilized
Pureté ou qualité > 95%